siRNA Details
Gene Symbol: RGD1561282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109213.1 | NP_001102683.1 | 2, 3 | 406 | ||
XM_008762537.1 | XP_008760759.1 | 3, 4 | 2867 | |||
XM_008762538.1 | XP_008760760.1 | 3, 4 | 2867 | |||
XM_008762539.1 | XP_008760761.1 | 3, 4 | 1588 |
