siRNA Details
Gene Symbol: RGD1561282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109213.1 | NP_001102683.1 | 3 | 535 | ||
XM_008762534.1 | XP_008760756.1 | 3 | 3089 | |||
XM_008762535.1 | XP_008760757.1 | 3 | 3089 | |||
XM_008762536.1 | XP_008760758.1 | 3 | 1810 | |||
XM_008762537.1 | XP_008760759.1 | 4 | 2996 | |||
XM_008762538.1 | XP_008760760.1 | 4 | 2996 | |||
XM_008762539.1 | XP_008760761.1 | 4 | 1717 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
