siRNA Details
Gene Symbol: Chp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024139.2 | NP_077053.1 | 6 | 562 | ||
GenBank mRNA | AB070350.1 | - | 6 | 547 | ||
BC062029.1 | - | 6 | 562 | 5623922 | ||
U39875.1 | - | 6 | 808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
