siRNA Details
Gene Symbol: Asb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109216.2 | NP_001102686.1 | 4, 5 | 1314 | ||
XM_006235959.2 | XP_006236021.1 | 4, 5 | 1350 | |||
GenBank mRNA | BC168867.1 | - | 4, 5 | 1314 | 7191440 | |
FQ215594.1 | - | 4, 5 | 1196 |
