siRNA Details
Gene Symbol: Nupl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109217.1 | NP_001102687.1 | 7 | 1189 | ||
GenBank mRNA | FQ227080.1 | - | 7 | 1156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
