siRNA Details
Gene Symbol: Fam185a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128188.1 | NP_001121660.1 | 8 | 1197 | ||
GenBank mRNA | BC161871.1 | - | 8 | 1197 | 7112030 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
