siRNA Details
Gene Symbol: Tmem243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109219.1 | NP_001102689.1 | 3, 4 | 424 | ||
XM_006236002.2 | XP_006236064.1 | 3, 4 | 256 | |||
XM_008762712.1 | XP_008760934.1 | 2, 3 | 442 | |||
XM_008762713.1 | XP_008760935.1 | 2, 3 | 718 | |||
GenBank mRNA | BC162037.1 | - | 2, 3 | 341 | 7099001 |
