siRNA Details
Gene Symbol: Tmem243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109219.1 | NP_001102689.1 | 4, 5 | 538 | ||
XM_008762712.1 | XP_008760934.1 | 3, 4 | 556 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081029, BC147128, AK076719, BC147127, BC147812, and others.
