siRNA Details
Gene Symbol: RGD1564345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077681.1 | NP_001071149.1 | 2 | 428 | ||
GenBank mRNA | BC127528.1 | - | 2 | 428 | 7114751 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1564345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077681.1 | NP_001071149.1 | 2 | 428 | ||
GenBank mRNA | BC127528.1 | - | 2 | 428 | 7114751 |