siRNA Details
Gene Symbol: Pex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109220.1 | NP_001102690.1 | 16 | 2703 | ||
XM_006236054.1 | XP_006236116.1 | 16 | 2737 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109220.1 | NP_001102690.1 | 16 | 2703 | ||
XM_006236054.1 | XP_006236116.1 | 16 | 2737 |