siRNA Details
Gene Symbol: LOC500035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047955.1 | NP_001041420.1 | 4 | 583 | ||
GenBank mRNA | AY539889.1 | - | 4 | 583 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
