siRNA Details
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236277.2 | XP_006236339.1 | 9, 10 | 3263 | ||
XM_006236278.2 | XP_006236340.1 | 8, 9 | 4852 | |||
XM_006236279.2 | XP_006236341.1 | 8, 9 | 4852 | |||
XM_008762844.1 | XP_008761066.1 | 8, 9 | 4854 | |||
XM_008762845.1 | XP_008761067.1 | 8, 9 | 4851 |

More Information
Other Species Targeted

Gene Ontology Categories:
