siRNA Details
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236277.2 | XP_006236339.1 | 7 | 2206 | ||
XM_006236278.2 | XP_006236340.1 | 6 | 3795 | |||
XM_006236279.2 | XP_006236341.1 | 6 | 3795 | |||
XM_008762844.1 | XP_008761066.1 | 6 | 3797 | |||
XM_008762845.1 | XP_008761067.1 | 6 | 3794 |
