siRNA Details
Gene Symbol: Fam131b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025046.1 | NP_001020217.2 | 4 | 336 | ||
GenBank mRNA | BC092642.1 | - | 4 | 336 | 7318030 |

More Information
Other Species Targeted

