siRNA Details
Gene Symbol: Fam131b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025046.1 | NP_001020217.2 | 5 | 524 | ||
XM_008762903.1 | XP_008761125.1 | 2 | 176 | |||
GenBank mRNA | BC092642.1 | - | 5 | 524 | 7318030 |
