siRNA Details
Gene Symbol: Lrrc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109231.1 | NP_001102701.1 | 3 | 495 | ||
XM_006236435.2 | XP_006236497.1 | 2 | 1512 | |||
XM_006236436.2 | XP_006236498.1 | 2 | 363 | |||
XM_006236437.2 | XP_006236499.1 | 2 | 530 | |||
XM_006236438.2 | XP_006236500.1 | 2 | 215 |
