siRNA Details
Gene Symbol: Gimap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024328.2 | NP_001019499.1 | 2 | 426 | ||
GenBank mRNA | AJ633682.1 | - | 865 | |||
BC091210.1 | - | 2 | 372 | 7381387 | ||
CV075794.1 | - | 2 | 426 | |||
DQ125348.1 | - | 2 | 449 | |||
DQ125349.1 | - | 2 | 449 | |||
FQ232730.1 | - | 2 | 457 |
