siRNA Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126296.2 | NP_001119768.2 | 6 | 871 | ||
XM_006236464.2 | XP_006236526.1 | 6 | 890 | |||
XM_006236465.2 | XP_006236527.1 | 6 | 890 | |||
XM_006236466.2 | XP_006236528.1 | 6 | 890 | |||
GenBank mRNA | BC161978.1 | - | 6 | 871 | 9029930 | |
FQ226359.1 | - | 1 | 27 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
