siRNA Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126296.2 | NP_001119768.2 | 2 | 323 | ||
XM_006236464.2 | XP_006236526.1 | 2 | 342 | |||
XM_006236465.2 | XP_006236527.1 | 2 | 342 | |||
XM_006236466.2 | XP_006236528.1 | 2 | 342 | |||
GenBank mRNA | BC161978.1 | - | 2 | 323 | 9029930 |

More Information
Other Species Targeted

Gene Ontology Categories:
