siRNA Details
Gene Symbol: Stk31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191971.1 | NP_001178900.1 | 9 | 1064 | ||
XM_008762906.1 | XP_008761128.1 | 9 | 1091 |

More Information
Other Species Targeted

Gene Ontology Categories:
