siRNA Details
Gene Symbol: LOC500124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024329.1 | NP_001019500.1 | 10 | 1423 | ||
GenBank mRNA | BC078992.1 | - | 10 | 1423 | 7112600 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC500124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024329.1 | NP_001019500.1 | 10 | 1423 | ||
GenBank mRNA | BC078992.1 | - | 10 | 1423 | 7112600 |