siRNA Details
Gene Symbol: Creb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134621.1 | NP_001128093.1 | 1690 | |||
XM_008762908.1 | XP_008761130.1 | 7 | 1445 | |||
XM_008762909.1 | XP_008761131.1 | 11 | 1840 |
