siRNA Details
Gene Symbol: Neurod6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109237.1 | NP_001102707.1 | 2 | 427 | ||
GenBank mRNA | BC168855.1 | - | 1 | 453 | 8365463 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Neurod6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109237.1 | NP_001102707.1 | 2 | 427 | ||
GenBank mRNA | BC168855.1 | - | 1 | 453 | 8365463 |