siRNA Details
Gene Symbol: Neurod6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109237.1 | NP_001102707.1 | 2 | 832 | ||
GenBank mRNA | BC168855.1 | - | 1 | 858 | 8365463 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009717, BC087831, AK144303, U29086, AK144360, and others.
Gene Ontology Categories:
