siRNA Details
Gene Symbol: Ptcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134718.1 | NP_001128190.1 | 14 | 1160 | ||
XM_008762995.1 | XP_008761217.1 | 17 | 1724 | |||
GenBank mRNA | BC098732.1 | - | 5 | 525 |
