siRNA Details
Gene Symbol: Atoh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109241.1 | NP_001102711.1 | 1 | 1006 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153778, BC023684, AB046528, AB049066, AB046527, and others.
Gene Ontology Categories:
