siRNA Details
Gene Symbol: Eva1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109243.1 | NP_001102713.1 | 3 | 985 | ||
XM_006236708.2 | XP_006236770.1 | 3 | 3078 | |||
XM_008763027.1 | XP_008761249.1 | 4 | 887 | |||
XM_008763028.1 | XP_008761250.1 | 3 | 1557 | |||
GenBank mRNA | FQ216129.1 | - | 3 | 935 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
