siRNA Details
Gene Symbol: Exoc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109246.1 | NP_001102716.1 | 14 | 1435 | ||
XM_008763075.1 | XP_008761297.1 | 14 | 1472 |

More Information
Other Species Targeted

Gene Ontology Categories:
