siRNA Details
Gene Symbol: Exoc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109246.1 | NP_001102716.1 | 14 | 1443 | ||
XM_008763075.1 | XP_008761297.1 | 14 | 1480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
