siRNA Details
Gene Symbol: LOC100910829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236799.2 | XP_006236861.1 | 3 | 892 | ||
XM_006236800.2 | XP_006236862.1 | 891 | ||||
XM_006236801.2 | XP_006236863.1 | 877 |
Gene Symbol: Cml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173449.1 | NP_001166920.1 | 1 | 620 | ||
XM_006236785.2 | XP_006236847.1 | 3 | 981 | |||
XM_006236786.2 | XP_006236848.1 | 3 | 1007 | |||
XM_006236787.2 | XP_006236849.1 | 3 | 991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
