siRNA Details
Gene Symbol: Aak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173450.1 | NP_001166921.1 | 17 | 2974 | ||
XM_006236833.2 | XP_006236895.1 | 18 | 2824 | |||
XM_006236834.2 | XP_006236896.1 | 18 | 2821 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001040106, NM_177762, BC043125, AK129272, BC141176, and others.
Gene Ontology Categories:
