siRNA Details
Gene Symbol: Arhgap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109247.1 | NP_001102717.1 | 10 | 1679 | ||
XM_006236838.2 | XP_006236900.1 | 10 | 3838 | |||
XM_006236839.2 | XP_006236901.1 | 9 | 1534 |

More Information
Other Species Targeted

Gene Ontology Categories:
