siRNA Details
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236864.2 | XP_006236926.1 | 5 | 1321 | ||
XM_008763079.1 | XP_008761301.1 | 5 | 1322 | |||
GenBank mRNA | BC168694.1 | - | 5 | 1284 | 7937199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
