siRNA Details
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030045.1 | NP_001025216.1 | 5, 6 | 968 | ||
GenBank mRNA | AY598952.1 | - | 5, 6 | 968 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030045.1 | NP_001025216.1 | 5, 6 | 968 | ||
GenBank mRNA | AY598952.1 | - | 5, 6 | 968 |