siRNA Details
Gene Symbol: Setmar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025048.1 | NP_001020219.1 | 2 | 318 | ||
XM_006236996.2 | XP_006237058.1 | 2 | 189 | |||
XM_006236997.2 | XP_006237059.1 | 2 | 413 | |||
XM_008763195.1 | XP_008761417.1 | 2 | 352 | |||
GenBank mRNA | BC088181.1 | - | 2 | 318 | 7364937 |
