siRNA Details
Gene Symbol: Fancd2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109251.1 | NP_001102721.1 | 3 | 498 | ||
XM_006237051.2 | XP_006237113.1 | 2 | 570 | |||
XM_006237052.2 | XP_006237114.1 | 2 | 1171 |

More Information
Other Species Targeted

