siRNA Details
Gene Symbol: Dcp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749821.3 | XP_003749869.1 | 9 | 2024 | ||
XM_008763272.1 | XP_008761494.1 | 9 | 2192 | |||
XM_575654.6 | XP_575654.3 | 9 | 2123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
