siRNA Details
Gene Symbol: Cecr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191977.1 | NP_001178906.1 | 14 | 1778 | ||
XM_008763251.1 | XP_008761473.1 | 14 | 2054 | |||
XM_008763252.1 | XP_008761474.1 | 14 | 2054 | |||
XM_008763253.1 | XP_008761475.1 | 13 | 1970 |
