siRNA Details
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024335.1 | NP_001019506.1 | 3 | 496 | ||
XM_006237380.2 | XP_006237442.1 | 3 | 2006 | |||
XM_006237381.2 | XP_006237443.1 | 3 | 2006 | |||
XM_006237382.2 | XP_006237444.1 | 3 | 2006 | |||
XM_008763293.1 | XP_008761515.1 | 4 | 1801 | |||
GenBank mRNA | BC095844.1 | - | 3 | 496 | 7389613 |
