siRNA Details
Gene Symbol: LOC500331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024337.1 | NP_001019508.1 | 4, 5 | 819 | ||
XM_006237415.2 | XP_006237477.1 | 3, 4 | 518 | |||
XM_008763321.1 | XP_008761543.1 | 2, 3 | 710 | |||
XM_008763322.1 | XP_008761544.1 | 3, 4 | 606 | |||
GenBank mRNA | BC086997.1 | - | 4, 5 | 819 | 7131139 |
Gene Symbol: LOC100910725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237437.2 | XP_006237499.1 | 3, 4 | 390 | ||
XM_006237438.2 | XP_006237500.1 | 305 | ||||
XM_008763340.1 | XP_008761562.1 | 710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
