siRNA Details
Gene Symbol: Clec1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191978.1 | NP_001178907.1 | 5, 6 | 703 | ||
XM_006237088.2 | XP_006237150.1 | 5, 6 | 2279 | |||
XM_006237089.2 | XP_006237151.1 | 4, 5 | 1024 | |||
XM_006237090.2 | XP_006237152.1 | 4, 5 | 2181 | |||
GenBank mRNA | FQ227184.1 | - | 5, 6 | 772 | ||
FQ227324.1 | - | 5, 6 | 772 | |||
GU357482.1 | - | 5, 6 | 553 |
