siRNA Details
Gene Symbol: Klrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029908.1 | NP_001025079.1 | 4 | 365 | ||
XM_008763323.1 | XP_008761545.1 | 4 | 735 | |||
XM_008763324.1 | XP_008761546.1 | 4 | 735 | |||
GenBank mRNA | AY197774.1 | - | 4 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
