siRNA Details
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763441.1 | XP_008761663.1 | 2 | 756 | ||
XM_008763442.1 | XP_008761664.1 | 4 | 744 | |||
XM_008763443.1 | XP_008761665.1 | 5 | 1499 |
