siRNA Details
Gene Symbol: Far2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237697.2 | XP_006237759.1 | 8, 9 | 2065 | ||
XM_575726.6 | XP_575726.2 | 8, 9 | 1362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
