siRNA Details
Gene Symbol: Tmem70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109258.1 | NP_001102728.1 | 3 | 469 | ||
GenBank mRNA | FQ213707.1 | - | 3 | 379 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109258.1 | NP_001102728.1 | 3 | 469 | ||
GenBank mRNA | FQ213707.1 | - | 3 | 379 |