siRNA Details
Gene Symbol: RGD1564053 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109259.2 | NP_001102729.2 | 1, 2 | 530 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001160371, NM_177173, NM_001160370, NM_001160369, BB649073, and others.
