siRNA Details
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024340.1 | NP_001019511.2 | 7 | 657 | ||
XM_006237781.2 | XP_006237843.1 | 6 | 3483 | |||
XM_006237782.2 | XP_006237844.1 | 7 | 1286 | |||
XM_006237784.2 | XP_006237846.1 | 7 | 3038 | |||
XM_006237785.2 | XP_006237847.1 | 6 | 3485 | |||
XM_006237787.2 | XP_006237849.1 | 6 | 3482 | |||
XM_006237788.2 | XP_006237850.1 | 6 | 3480 | |||
XM_006237789.2 | XP_006237851.1 | 6 | 3480 | |||
GenBank mRNA | BC083882.1 | - | 7 | 657 | 7131459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
