siRNA Details
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024340.1 | NP_001019511.2 | 14 | 1890 | ||
XM_006237781.2 | XP_006237843.1 | 13 | 4716 | |||
XM_006237782.2 | XP_006237844.1 | 14 | 2519 | |||
XM_006237784.2 | XP_006237846.1 | 14 | 4271 | |||
XM_006237785.2 | XP_006237847.1 | 13 | 4703 | |||
XM_006237789.2 | XP_006237851.1 | 12 | 4570 | |||
GenBank mRNA | BC083882.1 | - | 14 | 1890 | 7131459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
