siRNA Details
Gene Symbol: Mcmdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024340.1 | NP_001019511.2 | 6, 7 | 640 | ||
XM_006237781.2 | XP_006237843.1 | 5, 6 | 3466 | |||
XM_006237782.2 | XP_006237844.1 | 6, 7 | 1269 | |||
XM_006237784.2 | XP_006237846.1 | 6, 7 | 3021 | |||
XM_006237785.2 | XP_006237847.1 | 5, 6 | 3468 | |||
XM_006237787.2 | XP_006237849.1 | 5, 6 | 3465 | |||
XM_006237788.2 | XP_006237850.1 | 5, 6 | 3463 | |||
XM_006237789.2 | XP_006237851.1 | 5, 6 | 3463 | |||
GenBank mRNA | BC083882.1 | - | 6, 7 | 640 | 7131459 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177722, CA978963, AK031395.
Gene Ontology Categories:
