siRNA Details
Gene Symbol: Sit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024344.1 | NP_001019515.1 | 3, 4 | 363 | ||
GenBank mRNA | BC088199.1 | - | 3, 4 | 363 | 7377221 | |
FQ225852.1 | - | 2, 3 | 278 | |||
FQ233921.1 | - | 2, 3 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
